Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177103.4 | NP_796077.2 | 1 - 2 | 296 | 145 |
GenBank mRNA | AK042087.1 | - | 1 - 2 | 188 | 145 |
AK043171.1 | - | 1 - 2 | 181 | 145 | |
AK158338.1 | - | 1 - 2 | 161 | 145 | |
AY522918.1 | - | 1 - 2 | 160 | 145 | |
BC067014.1 | - | 1 - 2 | 63 | 145 |