Assay Design



Gene Transcripts

Gene Symbol
Cryzl1
Entrez Gene ID
66609
Gene Name
crystallin, zeta (quinone reductase)-like 1
Gene Aliases
2210407J23Rik, 2410006O11Rik, QOH-1
Chromosome Location
Chr.16: 91689322 - 91728802 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026994.1 NP_081270.1 4 - 5 473 97
GenBank mRNA AK017995.1 - 4 - 5 474 97
BY717916.1 - 4 - 5 474 97

View Full Product Details