Assay Design



Gene Transcripts

Gene Symbol
Slc5a2
Entrez Gene ID
246787
Gene Name
solute carrier family 5 (sodium/glucose cotransporter), member 2
Gene Aliases
Sglt2
Chromosome Location
Chr.7: 128265697 - 128272433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133254.3 NP_573517.1 4 - 5 458 75
GenBank mRNA AK128900.1 - 4 - 5 472 75
AY033886.1 - 4 - 5 468 75
BC022226.1 - 4 - 5 458 75

View Full Product Details