Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126338.2 | NP_001119810.1 | 2 - 3 | 629 | 82 |
NM_001278520.1 | NP_001265449.1 | 2 - 3 | 608 | 82 | |
GenBank mRNA | AF165165.1 | - | 2 - 3 | 608 | 82 |
AF165166.1 | - | 2 - 3 | 612 | 82 | |
AF192385.1 | - | 2 - 3 | 628 | 82 | |
BC025140.1 | - | 2 - 3 | 605 | 82 | |
DQ832281.1 | - | 2 - 3 | 630 | 82 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278259.1 | NP_001265188.1 | 4 - 5 | 851 | 82 |