Assay Design
Gene Transcripts
- Gene Symbol
- Ccl1
- Entrez Gene ID
-
20290
- Gene Name
- chemokine (C-C motif) ligand 1
- Gene Aliases
-
BF534335, I-309, Scya1, Tca-3
- Chromosome Location
-
Chr.11: 82176666 - 82179884 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC042744.1 |
- |
- |
1750 |
101 |
View Full Product Details