Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198095.2 | NP_932763.1 | 4 - 5 | 544 | 171 |
GenBank mRNA | AK009361.1 | - | 4 - 5 | 344 | 171 |
AK154399.1 | - | 4 - 5 | 537 | 171 | |
BC027328.1 | - | 4 - 5 | 578 | 171 | |
BC056638.1 | - | 4 - 5 | 551 | 171 | |
BC087949.1 | - | 4 - 5 | 536 | 171 |