Assay Design
Gene Transcripts
- Gene Symbol
- Maged2
- Entrez Gene ID
-
80884
- Gene Name
- melanoma antigen, family D, 2
- Gene Aliases
-
4833439A22Rik, Mage-d2, ORF1
- Chromosome Location
-
Chr.X: 150806421 - 150814340 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC023319.1 |
- |
1 - 2 |
301 |
167 |
View Full Product Details