Assay Design
Gene Transcripts
- Gene Symbol
- Ighg
- Entrez Gene ID
-
380794
- Gene Name
- Immunoglobulin heavy chain (gamma polypeptide)
- Gene Aliases
-
AU044919, IGHG2A
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB097847.1 |
- |
1 - 2 |
85 |
139 |
View Full Product Details