Assay Design
Gene Transcripts
- Gene Symbol
- Atp5o
- Entrez Gene ID
-
28080
- Gene Name
- ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
- Gene Aliases
-
ATPO, D12Wsu28e, OSCP
- Chromosome Location
-
Chr.16: 91925223 - 91931630 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138597.2 |
NP_613063.1 |
1 - 2 |
115 |
154 |
GenBank mRNA |
AK005309.1 |
- |
1 - 2 |
114 |
154 |
View Full Product Details