Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033875.2 | NP_001029047.1 | 1 - 2 | 138 | 157 |
GenBank mRNA | BC103715.1 | - | 1 - 2 | 136 | 157 |
BC115516.1 | - | 1 - 2 | 138 | 157 | |
BC115517.1 | - | 1 - 2 | 138 | 157 |