Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291894.1 | NP_001278823.1 | - | 1051 | 80 |
NM_013532.3 | NP_038560.1 | - | 1046 | 80 | |
GenBank mRNA | AK155750.1 | - | - | 1980 | 80 |
AK170368.1 | - | - | 932 | 80 | |
BC006591.1 | - | - | 947 | 80 | |
BC099532.1 | - | - | 938 | 80 | |
U05264.1 | - | - | 947 | 80 | |
U05266.1 | - | - | 830 | 80 |