Assay Design



Gene Transcripts

Gene Symbol
Imp3
Entrez Gene ID
102462
Gene Name
IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast)
Gene Aliases
1190002L16Rik, AI256594
Chromosome Location
Chr.9: 56937500 - 56938398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133976.2 NP_598737.1 1 - 1 569 142
GenBank mRNA AK004456.1 - 1 - 1 596 142
BC009145.1 - 1 - 1 572 142

View Full Product Details