Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104541.1 | NP_001098011.1 | 3 - 4 | 1309 | 151 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104540.1 | NP_001098010.1 | 3 - 4 | 1309 | 151 |