Assay Design
Gene Transcripts
- Gene Symbol
- Atp6v0c
- Entrez Gene ID
-
11984
- Gene Name
- ATPase, H+ transporting, lysosomal V0 subunit C
- Gene Aliases
-
Atp6c, Atp6c2, Atp6l, Atpl, Atpl-rs1, PL16, VATL, Vma3
- Chromosome Location
-
Chr.17: 24163866 - 24169429 on Build GRCm38
- Gene Symbol
- Atp6v0c-ps2
- Entrez Gene ID
-
100039636
- Gene Name
- ATPase, H+ transporting, lysosomal V0 subunit C, pseudogene 2
- Gene Aliases
-
Atp6l-ps2, Gm15487
- Chromosome Location
-
Chr.6: 37805975 - 37807107 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037854.1 |
|
1 - 1 |
190 |
145 |
View Full Product Details