Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016904.1 | NP_058600.1 | 1 - 2 | 141 | 123 |
GenBank mRNA | AB025409.1 | - | 1 - 2 | 141 | 123 |
AK004101.1 | - | 1 - 2 | 127 | 123 | |
AK010414.2 | - | 1 - 2 | 128 | 123 | |
AK011314.1 | - | 1 - 2 | 130 | 123 | |
AK011782.1 | - | 1 - 2 | 107 | 123 | |
BC076579.1 | - | 1 - 2 | 133 | 123 |