Assay Design



Gene Transcripts

Gene Symbol
Cycs
Entrez Gene ID
13063
Gene Name
cytochrome c, somatic
Gene Aliases
-
Chromosome Location
Chr.6: 50562563 - 50566474 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007808.4 NP_031834.1 3 - 3 1324 119
GenBank mRNA AK143887.1 - 3 - 3 1325 119

View Full Product Details