Assay Design
Gene Transcripts
- Gene Symbol
- H2-T22
- Entrez Gene ID
-
15039
- Gene Name
- histocompatibility 2, T region locus 22
- Gene Aliases
-
27b, H-2T17, H-2T22, H2-T17, T22b
- Chromosome Location
-
Chr.17: 36037133 - 36042742 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M35247.1 |
- |
1 - 2 |
338 |
81 |
View Full Product Details