Assay Design
Gene Transcripts
- Gene Symbol
- Evpl
- Entrez Gene ID
-
14027
- Gene Name
- envoplakin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 116220559 - 116238147 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_025276.3 |
NP_079552.3 |
6 - 7 |
742 |
72 |
View Full Product Details