Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001115130.1 | NP_001108602.1 | 6 - 6 | 5999 | 75 |
NM_172765.3 | NP_766353.2 | 6 - 6 | 5945 | 75 | |
GenBank mRNA | AK053995.1 | - | 1 - 1 | 1424 | 75 |
AK153955.1 | - | 1 - 1 | 1126 | 75 | |
BC090965.1 | - | 1 - 1 | 606 | 75 |