Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025507.2 | NP_079783.2 | 11 - 12 | 1165 | 164 |
GenBank mRNA | AK009218.1 | - | 6 - 7 | 520 | 164 |
AK012384.1 | - | 6 - 7 | 520 | 164 | |
AX775977.1 | - | 11 - 12 | 1140 | 164 | |
AX775979.1 | - | 11 - 12 | 1140 | 164 |