Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194444.2 | NP_919426.2 | 5 - 6 | 418 | 116 |
NM_194446.2 | NP_919428.1 | 5 - 6 | 456 | 116 | |
GenBank mRNA | AK133918.1 | - | 5 - 6 | 411 | 116 |
AK144801.1 | - | 5 - 6 | 428 | 116 | |
BC017131.1 | - | 5 - 6 | 393 | 116 | |
BC023736.1 | - | 5 - 6 | 437 | 116 | |
BC052096.1 | - | 4 - 5 | 322 | 116 | |
DQ288857.1 | - | 5 - 6 | 455 | 116 |