Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010771.6 | NP_034901.2 | 15 - 15 | 3369 | 164 |
GenBank mRNA | AK029510.1 | - | 14 - 14 | 2749 | 164 |
AK084822.1 | - | 1 - 1 | 189 | 164 | |
AK087939.1 | - | 15 - 15 | 3254 | 164 | |
AK088351.1 | - | 15 - 15 | 3263 | 164 | |
AK129199.1 | - | 15 - 15 | 3320 | 164 | |
AK135937.1 | - | 14 - 14 | 3099 | 164 | |
AK139050.1 | - | 1 - 1 | 191 | 164 | |
AK148059.1 | - | 1 - 1 | 61 | 164 | |
AK148384.1 | - | 8 - 8 | 1696 | 164 | |
AK157744.1 | - | 14 - 14 | 2465 | 164 | |
AK161128.1 | - | 15 - 15 | 3268 | 164 | |
BC090833.1 | - | 14 - 14 | 3142 | 164 |