Assay Design



Gene Transcripts

Gene Symbol
Tmem60
Entrez Gene ID
212090
Gene Name
transmembrane protein 60
Gene Aliases
BC027828
Chromosome Location
Chr.5: 20882453 - 20886870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177601.3 NP_808269.1 1 - 2 207 131
GenBank mRNA AK041036.1 - 1 - 2 179 131
CN834017.1 - 1 - 2 473 131

View Full Product Details