Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005525.2 | NP_001005525.1 | 7 - 8 | 921 | 129 |
GenBank mRNA | BC132550.1 | - | 5 - 6 | 705 | 129 |
BC132552.1 | - | 5 - 6 | 705 | 129 | |
DT902145.1 | - | 3 - 4 | 384 | 129 |