Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017983.2 | NP_001017983.2 | 2 - 3 | 655 | 106 |
NM_001168260.1 | NP_001161732.1 | 2 - 3 | 753 | 106 | |
GenBank mRNA | AK079870.1 | - | 1 - 2 | 1100 | 106 |
AK140027.1 | - | 2 - 3 | 615 | 106 | |
AK170795.1 | - | 2 - 3 | 708 | 106 | |
BC086662.1 | - | 2 - 3 | 585 | 106 |