Assay Design
Gene Transcripts
- Gene Symbol
- Chst9
- Entrez Gene ID
-
71367
- Gene Name
- carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9
- Gene Aliases
-
5430438D01Rik
- Chromosome Location
-
Chr.18: 15451773 - 15760415 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199055.2 |
NP_951010.1 |
3 - 4 |
212 |
83 |
GenBank mRNA |
AB106879.1 |
- |
3 - 4 |
212 |
83 |
View Full Product Details