Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271615.1 | NP_001258544.1 | 1 - 2 | 145 | 73 |
GenBank mRNA | AK042942.1 | - | 1 - 2 | 94 | 73 |
BC027295.1 | - | 1 - 2 | 125 | 73 | |
BY084492.1 | - | 1 - 2 | 146 | 73 |