Assay Design



Gene Transcripts

Gene Symbol
Vmn2r51
Entrez Gene ID
100042921
Gene Name
vomeronasal 2, receptor 51
Gene Aliases
ENSMUSG00000058685
Chromosome Location
Chr.7: 10087198 - 10105659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105179.1 NP_001098649.1 2 - 3 487 150

View Full Product Details