Assay Design



Gene Transcripts

Gene Symbol
Rhox2d
Entrez Gene ID
434760
Gene Name
reproductive homeobox 2D
Gene Aliases
EG434760, Rhox2.4, Rhox2c
Chromosome Location
Chr.X: 37493410 - 37497715 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081669.2 NP_001075138.2 1 - 2 102 105

View Full Product Details