Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159407.1 | NP_001152879.1 | 6 - 6 | 2087 | 125 |
NM_001159408.1 | NP_001152880.1 | 5 - 5 | 1884 | 125 | |
NM_054052.3 | NP_473393.2 | 4 - 4 | 1763 | 125 | |
GenBank mRNA | AB059620.1 | - | 1 - 1 | 936 | 125 |
AK032911.1 | - | 4 - 4 | 1677 | 125 | |
AK037166.1 | - | 5 - 5 | 1911 | 125 | |
AK048010.1 | - | 4 - 4 | 1648 | 125 | |
AK083839.1 | - | 4 - 4 | 1763 | 125 | |
AY029203.1 | - | 3 - 3 | 1578 | 125 | |
BC063076.1 | - | 4 - 4 | 1599 | 125 |