Assay Design



Gene Transcripts

Gene Symbol
Amigo1
Entrez Gene ID
229715
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
Amigo, ali2
Chromosome Location
Chr.3: 108186289 - 108192286 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004293.2 NP_001004293.1 2 - 2 4981 149
NM_146137.3 NP_666249.2 2 - 2 5016 149
GenBank mRNA AK173118.1 - 2 - 2 4970 149

View Full Product Details