Assay Design
Gene Transcripts
- Gene Symbol
- Eif4ebp1
- Entrez Gene ID
-
13685
- Gene Name
- eukaryotic translation initiation factor 4E binding protein 1
- Gene Aliases
-
4e-bp1, AA959816, PHAS-I
- Chromosome Location
-
Chr.8: 27260327 - 27275656 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK013033.1 |
- |
- |
1830 |
132 |
View Full Product Details