Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104529.1 | NP_001097999.1 | 2 - 2 | 780 | 114 |
NM_001271766.1 | NP_001258695.1 | 2 - 2 | 719 | 114 | |
NM_022320.4 | NP_071715.3 | 2 - 2 | 1016 | 114 | |
GenBank mRNA | AF200349.1 | - | 2 - 2 | 701 | 114 |
AK034870.1 | - | 2 - 2 | 800 | 114 | |
AK036503.1 | - | 2 - 2 | 778 | 114 | |
AK089198.1 | - | 2 - 2 | 1034 | 114 | |
AK154203.1 | - | 2 - 2 | 703 | 115 | |
AK170995.1 | - | 2 - 2 | 720 | 114 | |
BC027429.1 | - | 2 - 2 | 653 | 114 |