Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019685.2 | NP_062659.1 | 9 - 9 | 1178 | 142 |
GenBank mRNA | AF100694.1 | - | 9 - 9 | 1217 | 142 |
AK012170.1 | - | 9 - 9 | 1123 | 142 | |
AK132783.1 | - | 9 - 9 | 1160 | 142 | |
AK146258.1 | - | 9 - 9 | 1171 | 142 | |
AK146374.1 | - | 9 - 9 | 1171 | 142 | |
AK156078.1 | - | 9 - 9 | 1148 | 142 | |
BC004718.1 | - | 9 - 9 | 1144 | 142 |