Assay Design



Gene Transcripts

Gene Symbol
Rnase11
Entrez Gene ID
497113
Gene Name
ribonuclease, RNase A family, 11 (non-active)
Gene Aliases
Raj1
Chromosome Location
Chr.14: 51048694 - 51055702 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011877.2 NP_001011877.1 1 - 1 597 73
GenBank mRNA BC141252.1 - 1 - 1 597 73

View Full Product Details