Assay Design
Gene Transcripts
- Gene Symbol
- Ott
- Entrez Gene ID
-
18422
- Gene Name
- ovary testis transcribed
- Gene Aliases
-
AV272316
- Chromosome Location
-
Chr.X: 149097952 - 149138152 on Build GRCm38
- Gene Symbol
- Gm15080
- Entrez Gene ID
-
666184
- Gene Name
- predicted gene 15080
- Gene Aliases
-
OTTMUSG00000018964, Ott-like
- Chromosome Location
-
Chr.X: 147992993 - 148033174 on Build GRCm38
- Gene Symbol
- Gm15085
- Entrez Gene ID
-
100039934
- Gene Name
- predicted gene 15085
- Gene Aliases
-
OTTMUSG00000018973
- Chromosome Location
-
Chr.X: 149447617 - 149487784 on Build GRCm38
- Gene Symbol
- Gm15093
- Entrez Gene ID
-
100039890
- Gene Name
- predicted gene 15093
- Gene Aliases
-
ENSMUSG00000072925, OTTMUSG00000018984
- Chromosome Location
-
Chr.X: 149272377 - 149305003 on Build GRCm38
- Gene Symbol
- Gm15107
- Entrez Gene ID
-
434864
- Gene Name
- predicted gene 15107
- Gene Aliases
-
OTTMUSG00000019001, Ott-like
- Chromosome Location
-
Chr.X: 148180675 - 148224710 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC109131.1 |
- |
- |
1355 |
143 |
- Gene Symbol
- Gm15114
- Entrez Gene ID
-
100034729
- Gene Name
- predicted gene 15114
- Gene Aliases
-
OTTMUSG00000019010, Ott-like
- Chromosome Location
-
Chr.X: 148488949 - 148521446 on Build GRCm38
View Full Product Details