Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012027.2 | NP_036157.2 | 19 - 20 | 3038 | 150 |
NM_201245.3 | NP_957697.1 | 19 - 20 | 3038 | 150 | |
GenBank mRNA | AB093269.1 | - | 7 - 8 | 4863 | 150 |
AK035435.1 | - | 13 - 14 | 2328 | 149 | |
AK154807.1 | - | 20 - 21 | 3045 | 150 | |
AK155630.1 | - | 19 - 20 | 2819 | 150 | |
AK158296.1 | - | 19 - 20 | 3027 | 150 | |
AK165710.1 | - | 4 - 5 | 992 | 150 | |
AK170849.1 | - | 19 - 20 | 3022 | 150 | |
AK171089.1 | - | 19 - 20 | 3027 | 150 | |
BC027417.1 | - | 5 - 6 | 1127 | 150 | |
BC039932.1 | - | 18 - 19 | 2562 | 150 | |
BC049803.1 | - | 19 - 20 | 2564 | 150 | |
BC063067.1 | - | 19 - 20 | 3001 | 150 | |
U73200.1 | - | 19 - 20 | 2898 | 150 |