Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004143.4 | NP_001004143.2 | 6 - 7 | 1016 | 171 |
GenBank mRNA | AK034492.1 | - | 6 - 7 | 991 | 171 |
AK084022.1 | - | 2 - 3 | 174 | 171 | |
AK155071.1 | - | 6 - 7 | 1017 | 171 | |
AK160992.1 | - | 6 - 7 | 925 | 171 | |
AK220368.1 | - | 6 - 7 | 977 | 171 | |
BC052540.1 | - | 2 - 3 | 311 | 171 | |
BC080737.1 | - | 6 - 7 | 887 | 171 |