Assay Design
Gene Transcripts
- Gene Symbol
- Olfr26
- Entrez Gene ID
-
18324
- Gene Name
- olfactory receptor 26
- Gene Aliases
-
MOR171-9, MTPCR09
- Chromosome Location
-
Chr.9: 38855060 - 38855990 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_146783.2 |
NP_666994.2 |
1 - 1 |
859 |
158 |
GenBank mRNA |
BC130238.1 |
- |
1 - 1 |
859 |
158 |
View Full Product Details