Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145568.3 | NP_663543.1 | 4 - 4 | 1629 | 148 |
GenBank mRNA | AK146818.1 | - | 3 - 3 | 1568 | 148 |
AK162678.1 | - | 3 - 3 | 1557 | 148 | |
BC005704.1 | - | 2 - 2 | 1274 | 148 | |
BC046798.1 | - | 3 - 3 | 1525 | 148 | |
BC069863.1 | - | 4 - 4 | 1629 | 148 |