Assay Design



Gene Transcripts

Gene Symbol
Nanog
Entrez Gene ID
71950
Gene Name
Nanog homeobox
Gene Aliases
2410002E02Rik, ENK, ecat4
Chromosome Location
Chr.6: 122706483 - 122714633 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289828.1 NP_001276757.1 2 - 3 627 90
NM_001289830.1 NP_001276759.1 3 - 4 424 90
NM_001289831.1 NP_001276760.1 3 - 4 408 90
NM_028016.3 NP_082292.1 2 - 3 627 90
GenBank mRNA AB126939.1 - 2 - 3 409 90
AF507043.1 - 2 - 3 599 90
AK010332.1 - 2 - 3 599 90
AK167259.1 - 2 - 3 598 90
AY278951.1 - 2 - 3 476 90
AY455282.1 - 3 - 4 408 90
AY455285.1 - 3 - 4 424 90
BC132017.1 - 2 - 3 624 90
BC137873.1 - 2 - 3 624 90
BC144995.1 - 2 - 3 624 90
BC144996.1 - 2 - 3 624 90

View Full Product Details