Assay Design



Gene Transcripts

Gene Symbol
Wnk4
Entrez Gene ID
69847
Gene Name
WNK lysine deficient protein kinase 4
Gene Aliases
2010002J11Rik, Pha2b, Prkwnk4
Chromosome Location
Chr.11: 101260537 - 101277409 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175638.3 NP_783569.1 9 - 10 2020 70
GenBank mRNA AY184228.1 - 9 - 10 1989 70
AY187027.1 - 9 - 10 1945 70
AY260059.1 - 9 - 10 1983 70
BC026679.1 - 2 - 3 219 70
BC037114.1 - 9 - 10 1980 70
BC043677.1 - 9 - 10 1393 70
BC096453.1 - 9 - 10 1976 70

View Full Product Details