Assay Design
Gene Transcripts
- Gene Symbol
- Tdpoz5
- Entrez Gene ID
-
399676
- Gene Name
- TD and POZ domain containing 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 93960292 - 93961314 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207273.2 |
NP_997156.2 |
- |
378 |
133 |
GenBank mRNA |
BC111104.1 |
- |
1 - 1 |
376 |
133 |
- Gene Symbol
- Gm10697
- Entrez Gene ID
-
100042761
- Gene Name
- predicted gene 10697
- Gene Aliases
-
ENSMUSG00000074425
- Chromosome Location
-
Chr.3: 94071622 - 94072644 on Build GRCm38
View Full Product Details