Assay Design
Gene Transcripts
- Gene Symbol
- Speer4a
- Entrez Gene ID
-
75657
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 4a
- Gene Aliases
-
1700027N01Rik, SPEER-4A
- Chromosome Location
-
Chr.5: 26017738 - 26042295 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_029376.2 |
NP_083652.2 |
5 - 5 |
730 |
147 |
GenBank mRNA |
AK006439.1 |
- |
5 - 5 |
734 |
147 |
View Full Product Details