Assay Design



Gene Transcripts

Gene Symbol
Cd59b
Entrez Gene ID
333883
Gene Name
CD59b antigen
Gene Aliases
-
Chromosome Location
Chr.2: 104069780 - 104085790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181858.1 NP_862906.1 - 354 79
GenBank mRNA BC139203.1 - 5 - 5 401 79
BC145912.1 - 5 - 5 401 79
BC171969.1 - 5 - 5 401 79

View Full Product Details