Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153101.3 | NP_694741.2 | 2 - 2 | 393 | 101 |
GenBank mRNA | AK156116.1 | - | 2 - 2 | 399 | 101 |
AY042192.1 | - | - | 367 | 101 | |
DY245072.1 | - | 1 - 1 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172588.1 | NP_001166059.1 | - | 368 | 101 |
GenBank mRNA | BC064040.1 | - | - | 368 | 101 |