Assay Design



Gene Transcripts

Gene Symbol
Bcl2
Entrez Gene ID
12043
Gene Name
B cell leukemia/lymphoma 2
Gene Aliases
AW986256, Bcl-2, C430015F12Rik, D630044D05Rik, D830018M01Rik
Chromosome Location
Chr.1: 106538178 - 106714290 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177410.2 NP_803129.2 1 - 1 2047 76
GenBank mRNA AK077913.1 - 1 - 1 827 76
BC068988.1 - 1 - 1 935 76

View Full Product Details