Assay Design



Gene Transcripts

Gene Symbol
Nms
Entrez Gene ID
433292
Gene Name
neuromedin S
Gene Aliases
AB164466
Chromosome Location
Chr.1: 38939149 - 38950276 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011684.2 NP_001011684.3 10 - 10 828 109
GenBank mRNA AB164466.1 - 10 - 10 828 109
BQ175462.1 - 1 - 1 109

View Full Product Details