Assay Design



Gene Transcripts

Gene Symbol
Rhox1
Entrez Gene ID
385343
Gene Name
reproductive homeobox 1
Gene Aliases
-
Chromosome Location
Chr.X: 37213804 - 37222258 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025084.2 NP_001020255.2 3 - 4 589 66
GenBank mRNA DQ058638.1 - 3 - 4 589 66

View Full Product Details