Assay Design



Gene Transcripts

Gene Symbol
Glt6d1
Entrez Gene ID
71103
Gene Name
glycosyltransferase 6 domain containing 1
Gene Aliases
4933411C14Rik, GT6m7
Chromosome Location
Chr.2: 25771820 - 25815940 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039095.1 NP_001034184.1 4 - 5 325 68
GenBank mRNA AK016765.1 - 4 - 5 333 68
BC111823.1 - 4 - 5 325 68

View Full Product Details